Anti-HS6ST1 Mouse Monoclonal Antibody [clone: 9L832]
Catalog # USBIH7520
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:heparan sulfate 6-O-sulfotransferase 1
- Antigen symbol:HS6ST1
- Clonality:Monoclonal
- Clone:9L832
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:38 to 411
- Storage buffer:PBS, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 25 ?g/ml
Type: Primary
Antigen: HS6ST1 (heparan sulfate 6-O-sulfotransferase 1)
Clonality: monoclonal
Clone: 9L832
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human