Anti-HepA-Related Protein Rabbit Polyclonal Antibody
USBIH1887-50
- Antibody type:Primary
- Antigen name:HepA-Related Protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,01% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunochemistry: 5 ?g/ml; Western Blot: 1:500-1:1000; ELISA: 1:16000-1:64000
Type: Primary
Antigen: HepA-Related Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human