Anti-HSPD1 Mouse Monoclonal Antibody [clone: 7L8]
Catalog # USBIH1830-70C-200
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Heat shock 60kDa protein 1
- Antigen symbol:HSPD1
- Clonality:Monoclonal
- Clone:7L8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat,Mouse,Yeast,Hamster,Pig,Rabbit,Monkey,Chicken,Human,Bovine,Guinea pig,Dog,E. coli
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,1mM PMSF, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.25 ?g/ml
Type: Primary
Antigen: HSPD1 (heat shock 60kDa protein 1 (chaperonin))
Clonality: monoclonal
Clone: 7L8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Chicken, Dog, E. coli, Guinea pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Yeast