Anti-Hantavirus, glycoprotein 2 Mouse monoclonal antibody [clone: 5i51]
Catalog # USBIH1808-61
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Hantavirus, Glycoprotein 2
- Clonality:Monoclonal
- Clone:5i51
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western blot:Yes
- Form:liquid
- Amino acid number:1 to 219
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:10000; Western Blot: 1:16000
Type: Primary
Antigen: Hantavirus, glycoprotein 2
Clonality: monoclonal
Clone: 5i51
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: