Anti-G Protein-Coupled Receptor Family C Group 5 Member D, GPRC5D Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11D88]
Catalog # USBIG8603-46
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:G Protein Coupled Receptor Family C Group 5 Member D
- Antigen symbol:GPRC5D
- Clonality:Monoclonal
- Clone:11D88
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, 0,5% BSA and 0,1% sodium azide
- Storage temperature:Cool
- Concentration:~0,01 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: G Protein-Coupled Receptor Family C Group 5 Member D, GPRC5D
Clonality: monoclonal
Clone: 11D88
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human