Anti-G Protein-Coupled Receptor GPR80/GPR99/P2Y15 Rabbit polyclonal antibody
Catalog # USBIG8600-85T
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor GPR80/GPR99/P2Y15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 20 ?g/ml
Type: Primary
Antigen: G Protein-Coupled Receptor GPR80/GPR99/P2Y15
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human