Anti-G Protein Coupled Receptor 75 Mouse monoclonal antibody [clone: 11n81]
Catalog # USBIG8600-79L
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:G Protein Coupled Receptor 75
- Antigen symbol:GPR75
- Clonality:Monoclonal
- Clone:11n81
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:lyophilized powder
- Storage buffer:PBS,trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 8-25 ?g/ml
Type: Primary
Antigen: G Protein Coupled Receptor 75
Clonality: monoclonal
Clone: 11n81
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human