Anti-GCTM-5 Mouse monoclonal antibody [clone: 9H238]
Catalog # USBIG2023-60
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:GCTM-5
- Clonality:Monoclonal
- Clone:9H238
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 5-20 ?g/ml; Western Blot: 0.5-2.5 ?g/ml
Type: Primary
Antigen: GCTM-5
Clonality: monoclonal
Clone: 9H238
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human