Anti-FXY2 Goat Polyclonal Antibody
Catalog # USBIF9400-02B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:FXY2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Form:liquid
- Storage buffer:TBS, pH 7,3, 0,5% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: ELISA: 1:32000; Western Blot: 1:32000; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: FXY2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human