Anti-Fibronectin Mouse monoclonal antibody [clone: 9i67]
USBIF4215-20B
- Antibody type:Primary
- Antigen name:Fibronectin
- Antigen symbol:FN1
- Clonality:Monoclonal
- Clone:9i67
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Dog
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7.6, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Recommended dilution: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: Fibronectin (fibronectin)
Clonality: monoclonal
Clone: 9i67
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human