Anti-Fibroblast Surface Protein Mouse monoclonal antibody [clone: 8k221]
Catalog # USBIF4213-60
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Fibroblast Surface Protein
- Clonality:Monoclonal
- Clone:8k221
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgM
- Reactivity:Human
- Western blot:Yes
- Format:ascites
- Form:liquid
- Storage buffer:PBS, pH 7,4, 1% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:500; Immunoprecipitation: 1:500
Type: Primary
Antigen: Fibroblast Surface Protein
Clonality: monoclonal
Clone: 8k221
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human