Anti-FCRL4 Mouse monoclonal antibody Carboxyfluorescein [clone: 11C14]
USBIF0025-75A
- Antibody type:Primary
- Antigen name:Fc receptor-like 4
- Antigen symbol:FCRL4
- Clonality:Monoclonal
- Clone:11C14
- Conjugation:FAM (Carboxyfluorescein)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Amino acid number:16 to 385
- Storage buffer:sodium chloride, 0,5% BSA, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~0,05 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: FCRL4 (Fc receptor-like 4)
Clonality: monoclonal
Clone: 11C14
Conjugation: Carboxyfluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human