Anti-DNP Rabbit Polyclonal Antibody
Catalog # USBID8060-09-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Dinitrophenol
- Antigen symbol:DNP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% BSA.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.1-1.0 ?g/ml; Western Blot: 0.5-5 ?g/ml
Type: Primary
Antigen: DNP (Dinitrophenol)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: