Anti-Corticotropin Releasing Factor Rabbit Polyclonal Antibody
Catalog # USBIC7903-80Y
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Corticotropin releasing factor
- Antigen symbol:CRF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Format:Serum
- Form:lyophilized powder
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Radio Immunoassay
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Corticotropin Releasing Factor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat