Anti-ITGA4 Rat Monoclonal Antibody [clone: 9C10 (MFR.4.B)]
Catalog # USBIC2404-71A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha 4
- Antigen symbol:ITGA4
- Clonality:Monoclonal
- Clone:9C10 (MFR.4.B)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ITGA4 (integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor))
Clonality: Monoclonal
Clone: 9C10 (MFR.4.B)
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse