Anti-Carbohydrate Sulfotransferase Mouse monoclonal antibody unconjugated [clone: 11D5]
Catalog # USBIC1103-04F
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Carbohydrate Sulfotransferase
- Antigen symbol:CPS
- Clonality:Monoclonal
- Clone:11D5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Neutralisation:Yes
- Form:lyophilized powder
- Amino acid number:29 to 386
- Storage buffer:PBSwith 5% trehalose.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0.5-1 ?g/ml
Type: Primary
Antigen: Carbohydrate Sulfotransferase
Clonality: Monoclonal
Clone: 11D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human