Order Entry
Ireland
ContactUsLinkComponent
Anti-Calcineurin A, pan Rabbit polyclonal antibody unconjugated
Anti-Calcineurin A, pan Rabbit polyclonal antibody unconjugated
  USBIC0114-02
Anti-Calcineurin A, pan Rabbit polyclonal antibody unconjugated
  USBIC0114-02

 

  • Antibody type:
    Primary
  • Antigen name:
    Calcineurin A, pan
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Drosophila,
    Monkey,
    Human,
    Rat,
    Mouse
  • Form:
    liquid
  • Storage buffer:
    10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 100ug/ml BSA, 50% glycerol, 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: Western Blot: 1:1000; Immunoprecipitation:1:50; Immunofluorescence: 1:50; Flow Cytometry: 1:25

Type: Primary
Antigen: Calcineurin A, pan
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila, Human, Monkey, Mouse, Rat