Order Entry
Ireland
ContactUsLinkComponent
Anti-BPTF Mouse monoclonal antibody unconjugated [clone: 8C45]
Anti-BPTF Mouse monoclonal antibody unconjugated [clone: 8C45]
Catalog # USBIB2853-40
Supplier:  US Biological
Anti-BPTF Mouse monoclonal antibody unconjugated [clone: 8C45]
Catalog # USBIB2853-40
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    bromodomain PHD finger transcription factor
  • Antigen symbol:
    BPTF
  • Clonality:
    Monoclonal
  • Clone:
    8C45
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 1% BSA, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Tested applications:
    Dot Blot
  • Purification:
    purified by Protein G chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: BPTF (bromodomain PHD finger transcription factor)
Clonality: monoclonal
Clone: 8C45
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: