Anti-BAI2 Mouse monoclonal antibody unconjugated [clone: 9G321]
Catalog # USBIB2704-05D
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:brain-specific angiogenesis inhibitor 2
- Antigen symbol:BAI2
- Clonality:Monoclonal
- Clone:9G321
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:lyophilized powder
- Amino acid number:22 to 108
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: BAI2 (brain-specific angiogenesis inhibitor 2)
Clonality: monoclonal
Clone: 9G321
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human