Anti-BACE1 Rabbit Polyclonal Antibody
USBIB0002-91B1
- Antibody type:Primary
- Antigen name:beta-site APP-cleaving enzyme 1
- Antigen symbol:BACE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,02% sodium azide, 30% glycerol.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:40 µg
- Pk:40 µG
Recommended dilution: ELISA: 0. 05-0.2?g/ml; Western Blot: 0.1-1 ?g/ml
Type: Primary
Antigen: BACE1 (beta-site APP-cleaving enzyme 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human