Order Entry
Ireland
ContactUsLinkComponent
Anti-AMP Activated Protein Kinase alpha1/AMPKa2 Rabbit Polyclonal Antibody
Anti-AMP Activated Protein Kinase alpha1/AMPKa2 Rabbit Polyclonal Antibody
Catalog # USBIA1475-05B
Supplier:  US Biological
Anti-AMP Activated Protein Kinase alpha1/AMPKa2 Rabbit Polyclonal Antibody
Catalog # USBIA1475-05B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AMP Activated Protein Kinase alpha 1/2
  • Antigen symbol:
    AMPKA1/2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Monkey,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Modification:
    phospho-Ser485, phospho-Ser491
  • Storage buffer:
    10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 100ug/ml BSA, 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

  • Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: AMP Activated Protein Kinase alpha1/AMPKa2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey, Mouse, Rat