Anti-ALDH3A1 Rabbit Polyclonal Antibody
Catalog # USBIA1334-33M2
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:aldehyde dehydrogenase 3 family, member A1
- Antigen symbol:ALDH3A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Storage buffer:PBS, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:50-1:100; ELISA: 1:1000; Immunochemistry: 1:50-1:100; Flow Cytometry: 1:10-1:50
Type: Primary
Antigen: ALDH3A1 (aldehyde dehydrogenase 3 family, member A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human