Order Entry
Ireland
ContactUsLinkComponent
Anti-ALDH3A1 Rabbit Polyclonal Antibody
Anti-ALDH3A1 Rabbit Polyclonal Antibody
  USBIA1334-33M
Anti-ALDH3A1 Rabbit Polyclonal Antibody
  USBIA1334-33M

 

  • Antibody type:
    Primary
  • Antigen name:
    aldehyde dehydrogenase 3 family, member A1
  • Antigen symbol:
    ALDH3A1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: ALDH3A1 (aldehyde dehydrogenase 3 family, member A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat