Anti-ALDH Rabbit Polyclonal Antibody
USBIA1334-03A
- Antibody type:Primary
- Antigen name:Aldehyde Dehydrogenase
- Antigen symbol:ALDH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,2, dextran. 500ul sterile ddH2O.Dilute further
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: ALDH (aldehyde dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: