Anti-A20 Zinc Finger Protein Mouse Monoclonal Antibody [clone: 6D88]
Catalog # USBIA0000-20
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:A20 Zinc Finger Protein
- Clonality:Monoclonal
- Clone:6D88
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:375 to 537
- Storage buffer:PBS containing 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: A20 Zinc Finger Protein
Clonality: monoclonal
Clone: 6D88
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human