Anti-MS4A4A Mouse Monoclonal Antibody [clone: 818112]
USBI145529
- Antibody type:Primary
- Antigen name:Membrane-spanning 4-Domains, Subfamily A, Member 4A
- Antigen symbol:MS4A4A
- Clonality:Monoclonal
- Clone:818112
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS,Trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: MS4A4A (membrane-spanning 4-domains, subfamily A, member 4A)
Clonality: monoclonal
Clone: 818112
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human