Anti-TPH1 Mouse Monoclonal Antibody [clone: Try-63]
USBI145333
- Antibody type:Primary
- Antigen name:tryptophan hydroxylase 1
- Antigen symbol:TPH1
- Clonality:Monoclonal
- Clone:Try-63
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG3
- Reactivity:Rabbit,Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Format:ascites
- Form:lyophilized powder
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: TPH1 (tryptophan hydroxylase 1)
Clonality: monoclonal
Clone: Try-63
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Rabbit, Rat