Anti-TMSB10 Mouse Monoclonal Antibody [clone: 2E3]
Catalog # USBI134528
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:thymosin beta 10
- Antigen symbol:TMSB10
- Clonality:Monoclonal
- Clone:2E3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MADKPDMGEIASFDKAKLKKTETQEKNTLPTKETIEQEKRSEIS
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TMSB10 (thymosin beta 10)
Clonality: monoclonal
Clone: 2000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human