Anti-PTGIR Mouse Monoclonal Antibody [clone: 4B10]
USBI132030
- Antibody type:Primary
- Antigen name:prostaglandin I2 (prostacyclin) receptor (IP)
- Antigen symbol:PTGIR
- Clonality:Monoclonal
- Clone:4B10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:296 to 387
- Storage buffer:PBS, pH 7,2.
- Sequence:RKAVFQRLKLWVCCLCLGPAHGDSQTPLSQLASGRRDPRAPSAPVGKEGSCVPLSAWGEGQVEPLPPTQQSSGSAVGTSSKAEASVACSLC
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunochemistry: 3?g/ml
Type: Primary
Antigen: PTGIR (prostaglandin I2 (prostacyclin) receptor (IP))
Clonality: monoclonal
Clone: 4B10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human