Anti-OAS1 Mouse Monoclonal Antibody [clone: 2D4]
Catalog # USBI130676
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:2'-5'-oligoadenylate synthetase 2, 69/71kDa
- Antigen symbol:OAS2
- Clonality:Monoclonal
- Clone:2D4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:SVSRRDKSKQVWEAVLLPLSLLSMMDLRNTPAKSLDKFIEDYLLPDTCFRMQINHAIDIICGFLKERCFRGSSYPVCVSKVVKGGSSGKGTTLRGRSDAD
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml
Type: Primary
Antigen: OAS1 (2'-5'-oligoadenylate synthetase 1, 40/46kDa)
Clonality: monoclonal
Clone: 2D4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human