Anti-IGDCC4 Mouse Monoclonal Antibody [clone: 6E6]
Catalog # USBI130469
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:immunoglobulin superfamily, DCC subclass, member 4
- Antigen symbol:IGDCC4
- Clonality:Monoclonal
- Clone:6E6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:1152 to 1251
- Storage buffer:PBS, pH 7,2.
- Sequence:DLEPEDPLPPEAPDLISGVGDPGQGAAWLDRELGGCELAAPGPDRLTCLPEAASASCSYPDLQPGEVLEETPGDSCQLKSPCPLGASPGLPRSPVSSSA
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: IGDCC4 (immunoglobulin superfamily, DCC subclass, member 4)
Clonality: monoclonal
Clone: 6000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human