Anti-MARCH3 Mouse Monoclonal Antibody [clone: 1F6]
Catalog # USBI129429
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Membrane-associated ring finger (C3HC4) 7, E3 ubiquitin protein ligase
- Antigen symbol:MARCH7
- Clonality:Monoclonal
- Clone:1F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Antigen synonyms:AXOT|RNF177|AXO|MARCH-VII
- Amino acid number:1 to 101
- Storage buffer:PBS, pH 7,2.
- Sequence:MTTSRCSHLPEVLPDCTSSAAPVVKTVEDCGSLVNGQPQYVMQVSAKDGQLLSTVVRTLATQSPFNDRPMCRICHEGSSQEDLLSPCECTGTLGTIHRSC
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MARCH3 (membrane-associated ring finger (C3HC4) 3, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: 1F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human