Order Entry
Ireland
ContactUsLinkComponent
Anti-KCNF1 Mouse Monoclonal Antibody [clone: 1B2-7B6]
Anti-KCNF1 Mouse Monoclonal Antibody [clone: 1B2-7B6]
Catalog # USBI128739
Supplier:  US Biological
Anti-KCNF1 Mouse Monoclonal Antibody [clone: 1B2-7B6]
Catalog # USBI128739
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    potassium voltage-gated channel, subfamily F, member 1
  • Antigen symbol:
    KCNF1
  • Clonality:
    Monoclonal
  • Clone:
    1B2-7B6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2.
  • Sequence:
    MDGSGERSLPEPGSQSSAASDDIEIVVNVGGVRQVLYGDLLSQYPETRLAELINCLAGGYDTIFSLCDDYDPGKREFYFDRDPDAFKCVIEVYYFGEVHMKKGICPICFKNEMDFWKVDLKFLDDCCKSHLSEKREELEEIARRVQLILDDLGVDAAEGRWRRCQKCVWKFLEKPESSCPARVVAVLSFLLILVSSVVMCMGTIPELQVLDAEGNRVEHPTLENVETACIGWFTLEYLLRLFSSPNKLHFALSFMNIVDVLAILPFYVSLTLTHLGARMMELTNVQQAVQALRIMRIARIFKLARHSSGLQTLTYALKRSFKELGLLLMYLAVGIFVFSALGYTMEQSHPETLFKSIPQSFWWAIITMTTVGYGDIYPKTTLGKLNAAISFLCGVIAIALPIHPIINNFVRYYNKQRVLETAAKHELELMELNSSSGGEGKTGGSRSDLDNLPPEPAGKEAPSCSSRLKLSHSDTFIPLLTEEKHHRTRLQSCK
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: KCNF1 (potassium voltage-gated channel, subfamily F, member 1)
Clonality: monoclonal
Clone: 1B2-7B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human