Anti-ATRX Mouse Monoclonal Antibody [clone: 3C9]
Catalog # USBI123754
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:alpha thalassemia/mental retardation syndrome X-linked
- Antigen symbol:ATRX
- Clonality:Monoclonal
- Clone:3C9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:2311 to 2410
- Storage buffer:PBS, pH 7,2.
- Sequence:FNLGALSAMSNQQLEDLINQGREKVVEATNSVTAVRIQPLEDIISAVWKENMNLSEAQVQALALSRQASQELDVKRREAIYNDVLTKQQMLISCVQRILM
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ATRX (alpha thalassemia/mental retardation syndrome X-linked)
Clonality: monoclonal
Clone: 3C9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human