Anti-ALPK2 Mouse Monoclonal Antibody [clone: 4E5]
Catalog # USBI123253
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Alpha-Kinase 2
- Antigen symbol:ALPK2
- Clonality:Monoclonal
- Clone:4E5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:201 to 310
- Storage buffer:PBS, pH 7,2.
- Sequence:DGTSSVTEQGRYKLPTAPEAAENDYPGIQGETRDSHQAREEFASDNLLNMDESVRETEMKLLSGESENSGMSQCWETAADKRVGGKDLWSKRGSRKSARVRQPGMKGNP
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ALPK2 (alpha-kinase 2)
Clonality: monoclonal
Clone: 400000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human