Anti-Caspase-11, p20 Mouse Monoclonal Antibody [clone: Flamy-2]
Catalog # USBI044635
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Caspase 11, p20
- Antigen symbol:CASP11
- Clonality:Monoclonal
- Clone:Flamy-2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Western blot:Yes
- Form:liquid
- Amino acid number:81 to 373
- Storage buffer:PBS, 10% glycerol, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Caspase-11, p20
Clonality: Monoclonal
Clone: Flamy-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: