Anti-TBC1D13 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBI042629-BIOTIN
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:TBC1 domain family, member 13
- Antigen symbol:TBC1D13
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:internal
- Form:liquid
- Amino acid number:83 to 113
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50
Type: Primary
Antigen: TBC1D13 (TBC1 domain family, member 13)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse