Anti-MAP2 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBI038122-BIOTIN
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:microtubule-associated Protein 2
- Antigen symbol:MAP2
- Clonality:Polyclonal
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100; Flow Cytometry: 1:10-1:50
Type: Primary
Antigen: MAP2 (microtubule-associated protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human