Anti-MAP1LC3B Rabbit Polyclonal Antibody (Biotin)
Catalog # USBI038114-BIOTIN
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:microtubule-associated Protein 1 light chain 3 beta
- Antigen symbol:MAP1LC3B
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:liquid
- Modification:phospho-Thr93, phospho-Tyr99
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Dot Blot
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Dot blot 1:100-500
Type: Primary
Antigen: MAP1LC3B (microtubule-associated protein 1 light chain 3 beta)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human