Anti-Dok5, PTB domain Rabbit Polyclonal Antibody
Catalog # USBI034764
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Docking Protein 5
- Antigen symbol:DOK5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by saturated ammonium sulfate precipitation followed by dialysis against PBS
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100
Type: Primary
Antigen: Dok5, PTB domain
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human