Anti-TECRL Mouse Monoclonal Antibody [clone: 601CT22.4.3]
Catalog # USBI031265
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:trans-2,3-enoyl-CoA reductase-like
- Antigen symbol:TECRL
- Clonality:Monoclonal
- Clone:601CT22.4.3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Format:ascites
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-400
Type: Primary
Antigen: TECRL (trans-2,3-enoyl-CoA reductase-like)
Clonality: Monoclonal
Clone: 601CT22.4.3
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human