Anti-PRTN3 Mouse Monoclonal Antibody [clone: 13B647]
Catalog # USBI030569
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:proteinase 3
- Antigen symbol:PRTN3
- Clonality:Monoclonal
- Clone:13B647
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,03% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PRTN3 (proteinase 3)
Clonality: Monoclonal
Clone: 13B647
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human