Anti-PRH2 Mouse Monoclonal Antibody [clone: 13B639]
USBI030561
- Antibody type:Primary
- Antigen name:proline-rich Protein HaeIII subfamily 2
- Antigen symbol:PRH2
- Clonality:Monoclonal
- Clone:13B639
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Format:ascites
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: PRH2 (proline-rich protein HaeIII subfamily 2)
Clonality: Monoclonal
Clone: 13B639
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human