Anti-PPP1CA Mouse Monoclonal Antibody [clone: 13B636]
Catalog # USBI030558
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Protein phosphatase 1, catalytic subunit, alpha
- Antigen symbol:PPP1CA
- Clonality:Monoclonal
- Clone:13B636
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Format:ascites
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PPP1CA (protein phosphatase 1, catalytic subunit, alpha isozyme)
Clonality: Monoclonal
Clone: 13B636
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse