Anti-PHB Mouse Monoclonal Antibody [clone: 13B622]
USBI030544
- Antibody type:Primary
- Antigen name:prohibitin
- Antigen symbol:PHB
- Clonality:Monoclonal
- Clone:13B622
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Format:ascites
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: PHB (prohibitin)
Clonality: Monoclonal
Clone: 13B622
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat