Anti-PGR Mouse Monoclonal Antibody [clone: 13B620]
Catalog # USBI030542
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:progesterone receptor
- Antigen symbol:PGR
- Clonality:Monoclonal
- Clone:13B620
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Format:ascites
- Amino acid number:731 to 909
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PGR (progesterone receptor)
Clonality: Monoclonal
Clone: 13B620
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human