Anti-PBK Mouse Monoclonal Antibody [clone: 13B611]
Catalog # USBI030533
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:PDZ binding kinase
- Antigen symbol:PBK
- Clonality:Monoclonal
- Clone:13B611
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Format:ascites
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PBK (PDZ binding kinase)
Clonality: Monoclonal
Clone: 13B611
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human