Anti-NAMPT Mouse Monoclonal Antibody [clone: 13B610]
Catalog # USBI030532
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Nicotinamide phosphoribosyltransferase
- Antigen symbol:NAMPT
- Clonality:Monoclonal
- Clone:13B610
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Format:ascites
- Amino acid number:338 to 479
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: NAMPT (nicotinamide phosphoribosyltransferase)
Clonality: Monoclonal
Clone: 13B610
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human