Anti-Calcyclin Mouse Monoclonal Antibody [clone: 13B93]
Catalog # USBI030015
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Calcyclin
- Clonality:Monoclonal
- Clone:13B93
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Storage buffer:PBS, 0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Calcyclin
Clonality: Monoclonal
Clone: 13B93
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human