Anti-LSF Rabbit Polyclonal Antibody
USBI029530
- Antibody type:Primary
- Antigen symbol:LSF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:310 to 335
- Storage buffer:0,1M Tris-glycine, pH 7,4, 150mM sodium chloride, 0,05% sodium azide, 30% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:25 Tests
- Pk:25 Tests
Type: Primary
Antigen: LSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse